Thermo Scientific Chemicals

3-Chloro-4-fluoroaniline, 98%, Thermo Scientific Chemicals

Catalog number: 211530250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Chloro-4-fluoroaniline, 98%, Thermo Scientific Chemicals

Catalog number: 211530250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 211530250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
367-21-5
IUPAC Name
3-chloro-4-fluoroaniline
Molecular Formula
C6H5ClFN
InChI Key
YSEMCVGMNUUNRK-UHFFFAOYSA-N
SMILES
NC1=CC=C(F)C(Cl)=C1
Appearance (Color)
Beige to brown
Infrared spectrum
Conforms
Appearance
May darken on storage
Appearance (Form)
Crystalline solid
Melting point
42°C to 47°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text