Lead(II) nitrate, 99+%
Lead(II) nitrate, 99+%
Lead(II) nitrate, 99+%
Lead(II) nitrate, 99+%
Thermo Scientific Chemicals

Lead(II) nitrate, 99+%

CAS: 10099-74-8 | N2O6Pb | 331.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
1 kg
Packaging:
Plastic bottle
Catalog number 211560010
Price (USD)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS10099-74-8
IUPAC Nameλ²-lead(2+) dinitrate
Molecular FormulaN2O6Pb
InChI KeyRLJMLMKIBZAXJO-UHFFFAOYSA-N
SMILES[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(14g in 50ml H2O, 20°C) Clear colorless solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link