Thermo Scientific Chemicals

Magnesium acetate tetrahydrate, 97.5%, extra pure, crystalline, Thermo Scientific Chemicals

Catalog number: 211572500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Magnesium acetate tetrahydrate, 97.5%, extra pure, crystalline, Thermo Scientific Chemicals

Catalog number: 211572500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 211572500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16674-78-5
IUPAC Name
magnesium(2+) diacetate tetrahydrate
Molecular Formula
C4H14MgO8
InChI Key
XKPKPGCRSHFTKM-UHFFFAOYSA-L
SMILES
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals
Titration Complexometric
96.5 to 103.5 %
Sulfate (SO4)
=<500 ppm
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text