Sodium nitroferricyanide dihydrate, 99+%
Sodium nitroferricyanide dihydrate, 99+%
Sodium nitroferricyanide dihydrate, 99+%
Sodium nitroferricyanide dihydrate, 99+%
Thermo Scientific Chemicals

Sodium nitroferricyanide dihydrate, 99+%

CAS: 13755-38-9 | C5H4FeN6Na2O3 | 297.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 211640250
Price (USD)
55.90
Each
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
55.90
Each
Add to cart
Chemical Identifiers
CAS13755-38-9
IUPAC Namedisodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate
Molecular FormulaC5H4FeN6Na2O3
InChI KeyJFPDVRGGFBUORC-UHFFFAOYSA-N
SMILESO.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=99.0 %
Hexacyanoferrate III=<0.01 %
Appearance (Color)Red to dark red-brown
Sulfate (SO4)=<100 ppm
Hexacyanoferrate II=<0.02 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link