Thermo Scientific Chemicals

Sodium nitroferricyanide dihydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 211641000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Sodium nitroferricyanide dihydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 211641000
100 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 211641000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13755-38-9
IUPAC Name
disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate
Molecular Formula
C5H4FeN6Na2O3
InChI Key
JFPDVRGGFBUORC-UHFFFAOYSA-N
SMILES
O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
Appearance (Form)
Powder or crystals and/or chunks
Chloride (Cl)
=<100 ppm
Appearance (Color)
Red to dark red-brown
Sulfate (SO4)
=<100 ppm
Hexacyanoferrate II
=<0.02 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text