Zinc nitrate hexahydrate, 98%, extra pure, Thermo Scientific Chemicals
Zinc nitrate hexahydrate, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc nitrate hexahydrate, 98%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Packaging:
Plastic bottle
Catalog number 211660010
Price (USD)/ Each
125.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
148.00 
Save 22.35 (15%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
125.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
148.00 
Save 22.35 (15%)
Add to cart
Zinc nitrate hexahydrate, 98%, extra pure, Thermo Scientific Chemicals
Catalog number211660010
Price (USD)/ Each
125.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
148.00 
Save 22.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namezinc(2+) hexahydrate dinitrate
Molecular FormulaH12N2O12Zn
InChI KeyJGPSMWXKRPZZRG-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Molecular Weight (g/mol)297.48
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Adhering crystals and/or chunks
Titration Complexometric97.5 to 102.5 %
Iron (Fe)=<100 ppm
Lead (Pb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only