Thermo Scientific Chemicals

Zinc nitrate hexahydrate, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 211662500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Zinc nitrate hexahydrate, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 211662500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 211662500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10196-18-6
IUPAC Name
zinc(2+) hexahydrate dinitrate
Molecular Formula
H12N2O12Zn
InChI Key
JGPSMWXKRPZZRG-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
White
Appearance (Form)
Adhering crystals and/or chunks
Titration Complexometric
97.5 to 102.5 %
Iron (Fe)
=<100 ppm
Lead (Pb)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text