Thermo Scientific Chemicals

Isonicotinoyl chloride hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 211680250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Isonicotinoyl chloride hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 211680250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 211680250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
39178-35-3
IUPAC Name
hydrogen pyridine-4-carbonyl chloride chloride
Molecular Formula
C6H5Cl2NO
InChI Key
BNTRVUUJBGBGLZ-UHFFFAOYSA-N
SMILES
[H+].[Cl-].ClC(=O)C1=CC=NC=C1
Appearance (Form)
Crystalline powder
Melting point
153°C to 161°C
Appearance (Color)
White to beige
Infrared spectrum
Conforms
Titration Argentometric
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text