Isonicotinoyl chloride hydrochloride, 97%, Thermo Scientific Chemicals
Isonicotinoyl chloride hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isonicotinoyl chloride hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
Packaging:
Plastic bottle
Catalog number 211682500
Price (USD)/ Each
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS39178-35-3
IUPAC Namehydrogen pyridine-4-carbonyl chloride chloride
Molecular FormulaC6H5Cl2NO
InChI KeyBNTRVUUJBGBGLZ-UHFFFAOYSA-N
SMILES[H+].[Cl-].ClC(=O)C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point153°C to 161°C
Appearance (Color)White to beige
Infrared spectrumConforms
Titration Argentometric>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only