Bacto™ Proteose Peptone
Bacto™ Proteose Peptone
Gibco™

Bacto™ Proteose Peptone

Bacto Proteose Peptones are enzymatic digests of protein. Studies of peptic digests of animal tissue prepared under varying digestion parametersRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityForm
211684500 gDehydrated, free-flowing granules
21201010 kgDehydrated, free-flowing granules
2 Options
Catalog number 211684
Price (USD)/ 500 g
288.00
-
Add to cart
Quantity:
500 g
Form:
Dehydrated, free-flowing granules
Price (USD)/ 500 g
288.00
Add to cart
Bacto™ Proteose Peptone
Catalog number211684
Price (USD)/ 500 g
288.00
-
Add to cart
Bacto Proteose Peptones are enzymatic digests of protein. Studies of peptic digests of animal tissue prepared under varying digestion parameters demonstrated that no single peptone is the most suitable nitrogen source for every microbiological application. These findings led to the development of Bacto Proteose Peptone, Bacto Proteose Peptone No. 2, and Bacto Proteose Peptone No. 3. Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone.

• Bacto Proteose Peptone is used in preparing microbiological culture media and in producing bacterial toxins
• Bacto Proteose Peptone No. 2 is used in preparing microbiological culture media
• Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media
• Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone. It offers the same beneficial nutrients as Bacto Proteose Peptone for growth promotion and toxin production in a wide range of fastidious microorganisms.

We also offer alternatives to Bacto Proteose Peptones for scale-up to production applications, including BiTek Proteose Peptone.
Specifications
Certifications/ComplianceLUO, MUO
Cell TypeBacteria, Fungi, Mammalian
FormDehydrated, free-flowing granules
Product TypeBacterial Media
Shelf Life1825 Days
FormatBottle
For Use With (Application)Formulation of culture media for the cultivation of heterotrophic microorganisms
pH6.6–7.6 (1.0% solution at 25°C)
Product LineBacto™
Quantity500 g
SourceAnimal Origin
Unit Size500 g
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.