Kaolin, pure, Thermo Scientific Chemicals
Kaolin, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Kaolin, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 211740010
Price (USD)/ Each
94.65
Online exclusive
111.00 
Save 16.35 (15%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
94.65
Online exclusive
111.00 
Save 16.35 (15%)
Add to cart
Kaolin, pure, Thermo Scientific Chemicals
Catalog number211740010
Price (USD)/ Each
94.65
Online exclusive
111.00 
Save 16.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedioxo-2,4,6-trioxa-3,5-disila-1,7-dialuminaheptane-3,5-dione dihydrate
Molecular FormulaAl2H4O9Si2
InChI KeyNLYAJNPCOHFWQQ-UHFFFAOYSA-N
SMILESO.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O
Molecular Weight (g/mol)258.16
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light beige
Loss on drying=<1.5 %
Particle size50 to 62 % (< 2 µm)
Appearance (Form)Powder
Loss on ignition10.8 % to 11.8 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only