Kaolin, pure
Kaolin, pure
Kaolin, pure
Thermo Scientific Chemicals

Kaolin, pure

CAS: 1332-58-7 | Al2H4O9Si2 | 258.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 211740025
Price (EUR)
78,71
Online Exclusive
92,60
Save 13,89 (15%)
Each
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (EUR)
78,71
Online Exclusive
92,60
Save 13,89 (15%)
Each
Add to cart
Chemical Identifiers
CAS1332-58-7
IUPAC Namedioxo-2,4,6-trioxa-3,5-disila-1,7-dialuminaheptane-3,5-dione dihydrate
Molecular FormulaAl2H4O9Si2
InChI KeyNLYAJNPCOHFWQQ-UHFFFAOYSA-N
SMILESO.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Particle size50 to 62 % (< 2 µm)
Loss on ignition10.8 % to 11.8 %
Particle size1.0 to 1.8 µm (D50)
Appearance (Form)Powder
Appearance (Color)Light beige
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection