Appearance (Color)Yellow to golden to orange-brown
UV Lambda max409 to 415 nm (10 mg/l in water)
Appearance (Form)Powder
Solubility(1 mg/ml in methanol) Clear to slightly hazy yellow to yellow-green
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.