Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
100 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 211780010
Price (USD)/ Each
193.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
228.00 
Save 34.35 (15%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
193.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
228.00 
Save 34.35 (15%)
Add to cart
Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Catalog number211780010
Price (USD)/ Each
193.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
228.00 
Save 34.35 (15%)
-
Add to cart
Chemical Identifiers
CAS94-36-0
IUPAC Namebenzoyl benzenecarboperoxoate
Molecular FormulaC14H10O4
InChI KeyOMPJBNCRMGITSC-UHFFFAOYSA-N
SMILESO=C(OOC(=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Impurity=<0.25 % Inorganic and organic hydrolizable chloride
Appearance (Form)Moist granular powder
Titration Iodometric73 to 77 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiators
RUO – Research Use Only