Thermo Scientific Chemicals

L(+)-Tartaric acid diammonium salt, 99%, Thermo Scientific Chemicals

Catalog number: 211810010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

L(+)-Tartaric acid diammonium salt, 99%, Thermo Scientific Chemicals

Catalog number: 211810010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 211810010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3164-29-2
IUPAC Name
(2R,3R)-2,3-dihydroxybutanedioic acid diamine
Molecular Formula
C4H12N2O6
InChI Key
NGPGDYLVALNKEG-OLXYHTOASA-N
SMILES
N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O
Appearance (Color)
Semitransparent white
Infrared spectrum
Conforms
Sulfated ash
=<0.05 %
Chloride (Cl)
=<50 ppm
Insoluble matter
=<0.005 % (in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text