Thermo Scientific Chemicals

Cerium(III) chloride heptahydrate, 99%, Thermo Scientific Chemicals

Catalog number: 211820250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Cerium(III) chloride heptahydrate, 99%, Thermo Scientific Chemicals

Catalog number: 211820250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 211820250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18618-55-8
IUPAC Name
trichlorocerium heptahydrate
Molecular Formula
CeCl3H14O7
InChI Key
KPZSTOVTJYRDIO-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.O.Cl[Ce](Cl)Cl
Appearance (Form)
Crystalline powder or crystals
Titration Argentometric
>=98.5 %
Appearance (Color)
White to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text