Phenyl phosphate, disodium salt dihydrate, 98%, Thermo Scientific Chemicals
Phenyl phosphate, disodium salt dihydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenyl phosphate, disodium salt dihydrate, 98%, Thermo Scientific Chemicals

Phenyl phosphate, disodium salt dihydrate, 98%, C6H5Na2O4P.2H2O, CAS Number-66778-08-3, sodiumphenylphosphatedibasicdihydrate, 10g, White, 221-917-4, 254.09, HYGROSCOPIC, UMRLIPYPVZYYFT-UHFFFAOYSA-N, Authentic, phenyl dihydrogen phosphate;sodium;dihydrate, C6H5OP(O)(ONa)2.2H2O, >300.0 deg.C
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
Glass bottle
Catalog number 211830100
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
55.90 
Save 8.25 (15%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
55.90 
Save 8.25 (15%)
Add to cart
Phenyl phosphate, disodium salt dihydrate, 98%, Thermo Scientific Chemicals
Catalog number211830100
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
55.90 
Save 8.25 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namephenoxyphosphonic acid dihydrate disodium
Molecular FormulaC6H11Na2O6P
InChI KeyUMRLIPYPVZYYFT-UHFFFAOYSA-N
SMILESO.O.[Na].[Na].OP(O)(=O)OC1=CC=CC=C1
Molecular Weight (g/mol)256.10
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Water12 to 16 % (K.F.)
Appearance (Color)White to off-white
Titration with HClO4>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only