Thermo Scientific Chemicals

Phenyl phosphate, disodium salt dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 211831000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Phenyl phosphate, disodium salt dihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 211831000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 211831000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
66778-08-3
IUPAC Name
phenoxyphosphonic acid dihydrate disodium
Molecular Formula
C6H11Na2O6P
InChI Key
UMRLIPYPVZYYFT-UHFFFAOYSA-N
SMILES
O.O.[Na].[Na].OP(O)(=O)OC1=CC=CC=C1
Infrared spectrum
Conforms
Appearance (Color)
White to off-white
Titration with HClO4
>=97.5 %
Appearance (Form)
Crystalline powder
Water
12 to 16 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text