Thermo Scientific Chemicals

Ammonium iron(III) citrate, 16.5 to 18.5% Fe, Thermo Scientific Chemicals

Catalog number: 211840025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Ammonium iron(III) citrate, 16.5 to 18.5% Fe, Thermo Scientific Chemicals

Catalog number: 211840025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 211840025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1185-57-5
IUPAC Name
iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate amine
Molecular Formula
C6H8FeNO7
InChI Key
FRHBOQMZUOWXQL-UHFFFAOYSA-K
SMILES
N.[Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Appearance (Form)
Powder
Appearance (Color)
Brown-yellow to brown
Infrared spectrum
Conforms
Assay
16.5 to 18.5 % (Fe)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text