Ammonium iron(III) citrate, 16.5 to 18.5% Fe, Thermo Scientific Chemicals
Ammonium iron(III) citrate, 16.5 to 18.5% Fe, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium iron(III) citrate, 16.5 to 18.5% Fe, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 211842500
Price (USD)/ Each
71.65
Online exclusive
84.10 
Save 12.45 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
71.65
Online exclusive
84.10 
Save 12.45 (15%)
Add to cart
Ammonium iron(III) citrate, 16.5 to 18.5% Fe, Thermo Scientific Chemicals
Catalog number211842500
Price (USD)/ Each
71.65
Online exclusive
84.10 
Save 12.45 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameiron(3+) 2-hydroxypropane-1,2,3-tricarboxylate amine
Molecular FormulaC6H8FeNO7
InChI KeyFRHBOQMZUOWXQL-UHFFFAOYSA-K
SMILESN.[Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Molecular Weight (g/mol)261.98
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Brown-yellow to brown
Infrared spectrumConforms
Assay16.5 to 18.5 % (Fe)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only