Pierce™ Recombinant Protein A/G
Pierce™ Recombinant Protein A/G
Thermo Scientific™

Pierce™ Recombinant Protein A/G

Purified (unconjugated) Thermo Scientific Pierce Recombinant Protein A/G is useful as the basis for preparing various kinds of probes orRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
211865 mg
7767750 mg
77678500 mg
Catalog number 21186
Price (USD)
229.65
Online Exclusive
274.00
Save 44.35 (16%)
Each
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)
229.65
Online Exclusive
274.00
Save 44.35 (16%)
Each
Add to cart
Purified (unconjugated) Thermo Scientific Pierce Recombinant Protein A/G is useful as the basis for preparing various kinds of probes or affinity media for detection or purification of rabbit and human antibodies, especially IgG isotypes, in immunoassays and antibody purification protocols.

Features of Recombinant Protein A/G:

• Contains four Fc-binding domains from Protein A and two from Protein G
• Provides binding for all antibody species and subclasses recognized by either Protein A or Protein G

The extended Fc-binding properties of Protein A/G make it a popular tool in the investigation and purification of immunoglobulins. Protein A/G binds to all human IgG subclasses, IgA, IgE, IgM and to a lesser extent IgD; however, it does not bind mouse IgA, IgM or murine serum albumin. Protein A/G is an excellent tool for purification and detection of mouse monoclonal antibodies from IgG subclasses without interference from these other serum proteins. Individual subclasses of mouse monoclonals are most likely to have stronger affinity to this chimeric protein than to either Protein A or Protein G.

Properties of Recombinant Protein A/G:
• Source: E. coli
• Molecular weight: 50,460 (Apparent MW by SDS-PAGE: 40,000-45,000)
• Form: salt-free powder
• A280 of 0.1% solution: ∼0.505
• Isoelectric point (pI): 4.65

Gene fusion of the Fc-binding domains of Protein A and Protein G has resulted in production of a structurally and functionally chimeric protein with broader binding than either Protein A or Protein G alone. During fusion, the Protein G gene sequence coding for the serum albumin-binding site is eliminated. This protein is expressed in E. coli and secreted into the surrounding medium during fermentation. The product obtained is consistent in quality and yield because the bacterial host is engineered to be deficient in major proteolytic activities. Binding is less pH-dependent than either Protein A or Protein G alone, occurring well at pH 5 to 8.

Related Products
Pierce™ Recombinant Protein A/G, Alkaline Phosphatase Conjugated
Pierce™ Recombinant Protein A/G, Peroxidase Conjugated
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormPowder
Protein SubtypeProtein A
TargetAntibodies
Product LinePierce™
Product TypeProtein A/G
Quantity5 mg
Unit SizeEach
Contents & Storage
Upon receipt store at -20°C. Product is shipped at ambient temperature.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection