Benzyltrimethylammonium chloride, 98+%, Thermo Scientific Chemicals
Benzyltrimethylammonium chloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzyltrimethylammonium chloride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Packaging:
Glass bottle
Catalog number 211860010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namebenzyltrimethylazanium chloride
Molecular FormulaC10H16ClN
InChI KeyKXHPPCXNWTUNSB-UHFFFAOYSA-M
SMILES[Cl-].C[N+](C)(C)CC1=CC=CC=C1
Molecular Weight (g/mol)185.70
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=98.0 %
Appearance (Form)Crystalline powder and/or chunks
Appearance (Color)White to cream
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only