Yeast Extract
Yeast Extract
Gibco™

Yeast Extract

Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells. They are derived from primary grown baker’sRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2119291 lb.
2119305 lb.
21193125 lb.
Catalog number 211929
Price (USD)
177.00
Each
-
Add to cart
Quantity:
1 lb.
Price (USD)
177.00
Each
Add to cart
Ask our AI about this Product
Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells. They are derived from primary grown baker’s yeast. These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications. Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation.

Related yeast extract products:
• Bacto Yeast Extract—one of the most complete and versatile fermentation supplements available. It is an important ingredient for the microbiological assay of vitamins. Yeast extract is also of value in the assay of antibiotics. B factor, a growth substance necessary for the production of rifampin in a Nocardia species, can be isolated from yeast extract.
• Bacto Yeast Extract, Technical—developed to provide products priced for the biotechnology market with acceptable clarity and growth-promoting characteristics
• Gibco Yeast Extract—developed to provide a product for the biotechnology/pharmaceutical market with acceptable clarity and growth-promoting characteristics. Media formulations containing yeast extract are specified in standard methods for various applications.
• Difco Yeast Extract UF—ultra-filtered and specifically designed for mammalian cell culture applications. With its low endotoxin level and high content of naturally occurring B vitamins, it can be used effectively to reduce the need for fetal bovine serum. It has an endotoxin level of less than or equal to 500 EU/g.
• Difco Yeast Extract Low-Dusting (LD)—designed to limit the amount of dust generated compared to other spray-dried yeast extracts.
Specifications
Cell TypeBacteria
Certifications/ComplianceLUO
For Use With (Application)Formulation of culture media for the cultivation of heterotrophic microorganisms
FormatBottle
Quantity1 lb.
Shelf Life1460 Days
SourceAnimal origin-free
FormPowder
Product TypeBacterial Media
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

浏览此商品的顾客同时也查看了



Documents & Downloads

Certificates

Share catalog number, name or link

1x1 image pixel for data collection