Thermo Scientific Chemicals

Nuclear Fast Red, 94%, pure

Catalog number: 211980010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Nuclear Fast Red, 94%, pure

Catalog number: 211980010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 211980010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6409-77-4
IUPAC Name
sodium 4-amino-1,3-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate
Molecular Formula
C14H8NNaO7S
InChI Key
IFSXZLJQEKGQAF-UHFFFAOYSA-M
SMILES
[Na+].NC1=C(O)C(=C(O)C2=C1C(=O)C1=CC=CC=C1C2=O)S([O-])(=O)=O
Infrared spectrum
Conforms
Appearance (Form)
Powder
HPLC
>=92 %
Appearance (Color)
Dark red to bordeaux

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text