4-(Phenylazo)diphenylamine, 97%, Thermo Scientific Chemicals
4-(Phenylazo)diphenylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Phenylazo)diphenylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Packaging:
Glass bottle
Catalog number 212010250
Price (USD)/ Each
134.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
134.00
Add to cart
4-(Phenylazo)diphenylamine, 97%, Thermo Scientific Chemicals
Catalog number212010250
Price (USD)/ Each
134.00
-
Add to cart
Chemical Identifiers
CAS101-75-7
IUPAC NameN-phenyl-4-(2-phenyldiazen-1-yl)aniline
Molecular FormulaC18H15N3
InChI KeyVXLFYNFOITWQPM-UHFFFAOYSA-N
SMILESN(C1=CC=CC=C1)C1=CC=C(C=C1)N=NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point85°C to 91°C
HPLC>=96.0 %
Infrared spectrumConforms
Appearance (Color)Orange to brown
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only