Thermo Scientific Chemicals

Tri-n-butyltin methoxide, 97%, Thermo Scientific Chemicals

Catalog number: 212090100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Tri-n-butyltin methoxide, 97%, Thermo Scientific Chemicals

Catalog number: 212090100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 212090100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1067-52-3
IUPAC Name
1,1,1-tributyl-O-methylstannanol
Molecular Formula
C13H32OSn
InChI Key
SIIBSBQSPWMULI-UHFFFAOYSA-N
SMILES
CO.CCCC[SnH](CCCC)CCCC
Infrared spectrum
Conforms
Refractive index
1.4726 to 1.4746 (20°C, 589 nm)
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless
Elemental Analysis
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text