Tri-n-butyltin methoxide, 97%, Thermo Scientific Chemicals
Tri-n-butyltin methoxide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tri-n-butyltin methoxide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 212090250
Price (USD)/ Each
44.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
52.20 
Save 7.55 (14%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
44.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
52.20 
Save 7.55 (14%)
Add to cart
Tri-n-butyltin methoxide, 97%, Thermo Scientific Chemicals
Catalog number212090250
Price (USD)/ Each
44.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
52.20 
Save 7.55 (14%)
-
Add to cart
Chemical Identifiers
CAS1067-52-3
IUPAC Name1,1,1-tributyl-O-methylstannanol
Molecular FormulaC13H32OSn
InChI KeySIIBSBQSPWMULI-UHFFFAOYSA-N
SMILESCO.CCCC[SnH](CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4726 to 1.4746 (20°C, 589 nm)
Appearance (Form)Liquid
Elemental Analysis>=96.0 %
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only