Aluminum isopropoxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Aluminum isopropoxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum isopropoxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
View all
Glass bottle
Catalog number 212230100
Price (USD)/ Each
54.40
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
54.40
Add to cart
Aluminum isopropoxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
Catalog number212230100
Price (USD)/ Each
54.40
-
Add to cart
Chemical Identifiers
IUPAC Namealuminium(3+) tris(propan-2-olate)
Molecular FormulaC9H21AlO3
InChI KeySMZOGRDCAXLAAR-UHFFFAOYSA-N
SMILES[Al+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]
Molecular Weight (g/mol)204.25
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder and/or chunks
Total trace metal impurities=<150 ppm
Infrared spectrumConforms
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Lewis acid
RUO – Research Use Only