Aluminum nitrate nonahydrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Aluminum nitrate nonahydrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum nitrate nonahydrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
Packaging:
Glass bottle
Catalog number 212241000
Price (USD)/ Each
286.65
Online exclusive
337.00 
Save 50.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
286.65
Online exclusive
337.00 
Save 50.35 (15%)
Add to cart
Aluminum nitrate nonahydrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Catalog number212241000
Price (USD)/ Each
286.65
Online exclusive
337.00 
Save 50.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namealuminium(3+) nonahydrate trinitrate
Molecular FormulaAlH18N3O18
InChI KeySWCIQHXIXUMHKA-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Molecular Weight (g/mol)375.13
View more
SpecificationsSpecification SheetSpecification Sheet
Total trace metal impurities=<10 ppm
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration Complexometric6.5 to 7.9 % (Al)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only