Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure
Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure
Thermo Scientific™

Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure

Collect, store and transport dry chemicals and environmental samples with this large square wide-mouth HDPE bottle.
Have Questions?
Catalog number 2123-0010
Price (USD)/ Case of 6
160.65
Online exclusive
255.00 
Save 94.35 (37%)
-
Add to cart
Capacity (Metric):
4 L
Price (USD)/ Case of 6
160.65
Online exclusive
255.00 
Save 94.35 (37%)
Add to cart
Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure
Catalog number2123-0010
Price (USD)/ Case of 6
160.65
Online exclusive
255.00 
Save 94.35 (37%)
-
Add to cart

Collect, store and transport dry chemicals and environmental samples with Thermo Scientific™ Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure. Molded-in handgrips provide greater convenience and safety when pouring.

  • Useful for collecting, storing and transporting dry chemicals, environmental samples, reagents and distilled water
  • Good for freezing to -100°C (-148°F)
  • Excellent chemical resistance to most acids, bases and alcohols
  • Square shape saves space, handles easily
  • Wide mouth gives easy access
  • Polypropylene cap included
  • Not liquid leakproof
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Recycling Information:
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

Specifications
Capacity1 gal., 4 L
Capacity (Metric)4 L
Capacity (English)1 gal.
ColorTranslucent
DescriptionLarge Wide-Mouth Square Bottle
Diameter (English)5.6 in.
Diameter (English) Inner Neck3.5 in.
Diameter (Metric) Inner Neck89 mm
MaterialHDPE
No. per Pack1
SterilityNon-sterile
Closure ColorWhite
Certifications/ComplianceUSP Class VI; FDA - 21 CFR
Closure IncludedYes
GraduatedNo
MouthWide
ShapeSquare
Height (Metric)292 mm
Product TypeWide Mouth Bottle (Large)
Width (English)5.66 in.
Height (English)11.5 in.
Closure MaterialPolypropylene
Closure Size100 mm
Diameter (Metric)142 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizeCase of 6