Cadmium acetate dihydrate, 98%, pure, Thermo Scientific Chemicals
Cadmium acetate dihydrate, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium acetate dihydrate, 98%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 212312500
Price (USD)/ Each
57.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
67.80 
Save 10.15 (15%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
57.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
67.80 
Save 10.15 (15%)
Add to cart
Cadmium acetate dihydrate, 98%, pure, Thermo Scientific Chemicals
Catalog number212312500
Price (USD)/ Each
57.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
67.80 
Save 10.15 (15%)
-
Add to cart
Chemical Identifiers
CAS5743-04-4
IUPAC Namecadmium(2+) diacetate dihydrate
Molecular FormulaC4H10CdO6
InChI KeyAUIZLSZEDUYGDE-UHFFFAOYSA-L
SMILESO.O.[Cd++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystals
Titration Complexometric>=97.5 %
Loss on drying12.5 % to 14.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only