Thermo Scientific Chemicals

Cadmium nitrate tetrahydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 212342500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Cadmium nitrate tetrahydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 212342500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 212342500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10022-68-1
IUPAC Name
cadmium(2+) bis(nitric acid)
Molecular Formula
CdH2N2O6
InChI Key
IZIPRBSCGMISKX-UHFFFAOYSA-N
SMILES
[Cd++].O[N+]([O-])=O.O[N+]([O-])=O
Solubility
(10 % in water) Clear colorless
Chloride (Cl)
=<50 ppm
Iron (Fe)
=<20 ppm
Appearance (Form)
Crystalline powder or crystals
Insoluble matter
=<0.002 % (in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text