Thermo Scientific Chemicals

Ammonium cerium(IV) nitrate, pure, Thermo Scientific Chemicals

Catalog number: 212360010
1 kg, Each
Thermo Scientific Chemicals

Ammonium cerium(IV) nitrate, pure, Thermo Scientific Chemicals

Catalog number: 212360010
1 kg, Each
Quantity
Catalog number: 212360010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16774-21-3
IUPAC Name
λ⁴-cerium(4+) bis(nitric acid) diamine tetranitrate
Molecular Formula
CeH8N8O18
InChI Key
WIBGOERAEYJBOT-UHFFFAOYSA-N
SMILES
N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
Orange
Titration
>=98.5 % (On dry substance)
Appearance (Form)
Crystalline powder or crystals
Loss on drying
=<2 % (110°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text