Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), Thermo Scientific Chemicals
Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), Thermo Scientific Chemicals

Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), ACROS Organics, Quantity: 100g, Packaging: Glass bottle, CAS: 69365-72-6, White, Grade: Trace Metal Basis, Melting Point: 110.0 deg.C, Molecular Formula: GaN3O9.xH2O, Molecular Weight: 255.74, Percent Purity: 99.9998%
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 212440050
Price (USD)/ Each
57.65
Online exclusive
64.30 
Save 6.65 (10%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
57.65
Online exclusive
64.30 
Save 6.65 (10%)
Add to cart
Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), Thermo Scientific Chemicals
Catalog number212440050
Price (USD)/ Each
57.65
Online exclusive
64.30 
Save 6.65 (10%)
-
Add to cart
Chemical Identifiers
CAS69365-72-6
IUPAC Namegallium(3+) trinitrate
Molecular FormulaGaN3O9
InChI KeyCHPZKNULDCNCBW-UHFFFAOYSA-N
SMILES[Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Total trace metal impurities=<0.0002 %
Appearance (Color)White
Gallium (Ga)18 to 22 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only