Thermo Scientific Chemicals

Lithium bromide, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals

Catalog number: 212530250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Lithium bromide, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals

Catalog number: 212530250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 212530250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7550-35-8
IUPAC Name
lithium(1+) bromide
Molecular Formula
BrLi
InChI Key
AMXOYNBUYSYVKV-UHFFFAOYSA-M
SMILES
[Li+].[Br-]
Appearance (Form)
Powder or crystals and/or chunks
Total trace metal impurities
=<10 ppm
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text