Thermo Scientific Chemicals

Lithium nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 212540250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Lithium nitrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 212540250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 212540250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-69-4
IUPAC Name
lithium(1+) nitrate
Molecular Formula
LiNO3
InChI Key
IIPYXGDZVMZOAP-UHFFFAOYSA-N
SMILES
[Li+].[O-][N+]([O-])=O
Total trace metal impurities
=<10 ppm
Lithium (Li)
9 to 11 %
Appearance (Form)
Crystalline powder and/or chunks
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text