Thermo Scientific Chemicals

Potassium bromide, 99+%, for biochemistry

Catalog number: 212580010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Potassium bromide, 99+%, for biochemistry

Catalog number: 212580010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 212580010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7758-02-3
IUPAC Name
potassium bromide
Molecular Formula
BrK
InChI Key
IOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES
[K+].[Br-]
Insoluble matter
=<0.005 %
Sulfate (SO4)
=<0.005 %
Magnesium (Mg)
=<0.001 %
Iron (Fe)
=<0.0005 %
Sodium (Na)
=<0.02 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text