Potassium bromide, 99+%, for biochemistry
Potassium bromide, 99+%, for biochemistry
Thermo Scientific Chemicals

Potassium bromide, 99+%, for biochemistry

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 212582500
Price (USD)/ Each
94.10
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
94.10
Add to cart
Potassium bromide, 99+%, for biochemistry
Catalog number212582500
Price (USD)/ Each
94.10
-
Add to cart
Chemical Identifiers
CAS7758-02-3
IUPAC Namepotassium bromide
Molecular FormulaBrK
InChI KeyIOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES[K+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<0.005 %
Sulfate (SO4)=<0.005 %
Magnesium (Mg)=<0.001 %
Iron (Fe)=<0.0005 %
Sodium (Na)=<0.02 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
RUO – Research Use Only