Potassium phosphate, monobasic, 99+%, for biochemistry
Potassium phosphate, monobasic, 99+%, for biochemistry
Thermo Scientific Chemicals

Potassium phosphate, monobasic, 99+%, for biochemistry

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Catalog number 212590025
Price (USD)/ Each
-
Quantity:
2.5 kg
Request bulk or custom format
Chemical Identifiers
CAS7778-77-0
IUPAC Namepotassium dihydrogen phosphate
Molecular FormulaH2KO4P
InChI KeyGNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES[K+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
UVat 280 nm A: =<0.005
Appearance (Color)Colorless to white
UV(0.1 M in water)
Heavy metals (as Pb)=<0.001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only