Thermo Scientific Chemicals

Sodium iodide, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals

Catalog number: 212680010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Sodium iodide, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals

Catalog number: 212680010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 212680010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7681-82-5
IUPAC Name
sodium iodide
Molecular Formula
INa
InChI Key
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
SMILES
[Na+].[I-]
Appearance (Form)
Granular powder
Total trace metal impurities
=<0.001 %
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text