Thermo Scientific Chemicals

Potassium antimonyl tartrate sesquihydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 212801000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Potassium antimonyl tartrate sesquihydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 212801000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 212801000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
28300-74-5
IUPAC Name
dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate trihydrate
Molecular Formula
C8H10K2O15Sb2
InChI Key
WBTCZEPSIIFINA-UHFFFAOYNA-J
SMILES
O.O.O.[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2
Titration Iodometric
99.0 to 103.0 %
Titrable acid or base
=<0.020 meq/g
Appearance (Form)
Crystalline powder
Loss on drying
=<2.7 %
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text