Thermo Scientific Chemicals

Triethylsilane, 99%, Thermo Scientific Chemicals

Catalog number: 212920050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Triethylsilane, 99%, Thermo Scientific Chemicals

Catalog number: 212920050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 212920050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
617-86-7
IUPAC Name
triethylsilyl
Molecular Formula
C6H15Si
InChI Key
QXTIBZLKQPJVII-UHFFFAOYSA-N
SMILES
CC[Si](CC)CC
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Refractive index
1.4100 to 1.4120 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text