Thermo Scientific Chemicals

Diphenylphosphinyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 212960500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Diphenylphosphinyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 212960500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 212960500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1499-21-4
IUPAC Name
diphenylphosphinoyl chloride
Molecular Formula
C12H10ClOP
InChI Key
QPQGTZMAQRXCJW-UHFFFAOYSA-N
SMILES
ClP(=O)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Liquid
GC
>=97.5 %
Refractive index
1.6080 to 1.6120 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text