Thermo Scientific Chemicals

m-Anisoyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: 212970250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

m-Anisoyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: 212970250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 212970250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1711-05-3
IUPAC Name
3-methoxybenzoyl chloride
Molecular Formula
C8H7ClO2
InChI Key
RUQIUASLAXJZIE-UHFFFAOYSA-N
SMILES
COC1=CC=CC(=C1)C(Cl)=O
Infrared spectrum
Conforms
Refractive index
1.5575 to 1.5595 (20°C, 589 nm)
Titration RCOCl
>=98.5 %
Appearance (Color)
Clear colorless to light yellow
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text