DPBS, powder, calcium, magnesium
Save up to 29% on your order of cell culture products. View promotion details >
Product Image
Gibco™

DPBS, powder, calcium, magnesium

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washingRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2130002510 x 1 L
213000581 x 10 L
2130007450 L
Catalog number 21300025
Price (USD)
73.50
Each
-
Add to cart
Quantity:
10 x 1 L
Customize this product
Price (USD)
73.50
Each
Add to cart
Ask our AI about this Product

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

This DPBS is modified as follows:

  • With
    • Calcium
    • Magnesium
  • Without
    • Phenol Red

The complete formulation is available.

For in vitro diagnostic use. Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
Specifications
AdditiveNo Sodium Pyruvate
ClassificationAnimal Origin-Free
For Use With (Application)Mammalian Cell Culture
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality260 - 310 mOsm/kg
Phenol Red IndicatorNo Phenol Red
Product LineGibco™
Quantity10 x 1 L
Shipping ConditionRoom Temperature
FormPowder
Product TypeDPBS
Solution TypeDulbecco's Phosphate Buffered Saline
pH7.3 - 7.5
Unit SizeEach
Contents & Storage
Storage conditions: 15-30° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
four-plex-image-COS7-live-cell zoomed

Follow the 5 steps to publication-quality live-cell imaging.
Download your free guide today ›

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link