Thermo Scientific™

EZ-Link™ TFPA-PEG3-Biotin

Catalog number: 21303
Have Questions?
Thermo Scientific™

EZ-Link™ TFPA-PEG3-Biotin

Catalog number: 21303
Have Questions?
Catalog Number
21303
Unit Size
25 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
2130325 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific EZ-Link TFPA-PEG3-Biotin is an efficient, photoactivatable reagent based on tetrafluorophenyl azide for biotinylation and includes a 3-unit polyethylene glycol (PEG) spacer arm.

Features of EZ-Link TFPA-PEG3-Biotin:

Biomolecular labeling—biotinylate proteins, DNA, RNA and many other macromolecules, even if they do not possess primary amines or sulfhydryl groups
Photo-reactive—perfluorophenyl azido group activates upon exposure to ultraviolet light to form covalent bonds with nucleophiles and many other chemical groups
Pegylated—spacer arm contains a hydrophilic, 3-unit, polyethylene glycol (PEG) group
Enhances solubility—pegylation imparts water solubility to the biotinylated molecule, helping to prevent aggregation of biotinylated antibodies stored in solution
Irreversible—forms permanent thioether bonds; spacer arm cannot be cleaved
Solubility—best to dissolve in DMSO or DMF before further dilution in aqueous buffers
Long reach —spacer arm (total length added to target) is 33.4 angstroms, minimizing steric hindrance for binding interactions with streptavidin

TFPA-PEG3-Biotin is a photoactivatable reagent for biotinylation of antibodies, proteins and many other kinds of macromolecules. The tetrafluorophenyl azide (TFPA) group activates upon exposure to UV-Light (maximum absorptivity is at 320 nm) to insert covalently at sites containing C-H or N-H bonds. The hydrophilic polyethylene glycol (PEG) spacer arm imparts water solubility that is transferred to the biotinylated molecule, thus reducing aggregation of labeled molecules stored in solution. The PEG spacer arm also gives this reagent a long and flexible connection to minimize steric hindrance involved with binding to avidin molecules.

We manufacture biotin reagents to ensure the highest possible overall product integrity, consistency, and performance for the intended research applications.

Biotinylation reagents differ in reactivity, length, solubility, cell permeability and cleavability. Several different types of photoreactive compounds are available. Aryl azide reagents activate upon exposure to ultraviolet light initiate addition reactions with double bonds, insertion into C–H and N–H sites, or subsequent ring expansion to react with a nucleophile (e.g., primary amines).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
TFPA-PEG3-Biotin
Quantity
25 mg
Product Line
EZ-Link™
Cell Permeability
Cell-Permeant
Chemical Reactivity
Light-activated
Reactive Moiety
Aryl Azide
Solubility
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Label or Dye
Biotin
Spacer
Extra-long, Pegylated
Label Type
Biotin & Analogs

Contents & Storage

Store desiccated at 4°C. Shipped at ambient temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text