Thermo Scientific Chemicals

Methyl (S)-(-)-lactate, 97%, Thermo Scientific Chemicals

Catalog number: 213062500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl (S)-(-)-lactate, 97%, Thermo Scientific Chemicals

Catalog number: 213062500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 213062500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
27871-49-4
IUPAC Name
methyl (2S)-2-hydroxypropanoate
Molecular Formula
C4H8O3
InChI Key
LPEKGGXMPWTOCB-VKHMYHEASA-N
SMILES
COC(=O)[C@H](C)O
Appearance (Form)
Liquid
Free acid
=<1.0 % (lactic acid)
Refractive index
1.4120 to 1.4160 (20°C, 589 nm)
GC
>=96.0 %
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text