Thermo Scientific Chemicals

Cobalt(II) sulfate heptahydrate, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 213100025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Cobalt(II) sulfate heptahydrate, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 213100025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 213100025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10026-24-1
IUPAC Name
λ²-cobalt(2+) heptahydrate sulfate
Molecular Formula
CoH14O11S
InChI Key
MEYVLGVRTYSQHI-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O
Appearance (Form)
Adhering crystalline powder or crystals
Nickel (Ni)
=<500 ppm
Lead (Pb)
=<50 ppm
Titration Complexometric
>=99.0 %
Appearance (Color)
Red-brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text