Thermo Scientific Chemicals

Magnesium sulfate heptahydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 213110025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Magnesium sulfate heptahydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 213110025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 213110025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10034-99-8
IUPAC Name
magnesium(2+) heptahydrate sulfate
Molecular Formula
H14MgO11S
InChI Key
WRUGWIBCXHJTDG-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
Appearance (Color)
White
Titration Complexometric
>=99.4 %
pH
5 to 8 (5 % at 25°C)
Manganese (Mn)
=<5 ppm
Sodium (Na)
=<200 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text