Thermo Scientific Chemicals

Mercury(I) nitrate dihydrate, 98%, for analysis, Thermo Scientific Chemicals

Catalog number: 213132500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Mercury(I) nitrate dihydrate, 98%, for analysis, Thermo Scientific Chemicals

Catalog number: 213132500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 213132500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7782-86-7
IUPAC Name
mercury(1+) hydrate nitrate
Molecular Formula
H2HgNO4
InChI Key
UPCSQZXLTBTBCU-UHFFFAOYSA-N
SMILES
O.[Hg+].[O-][N+]([O-])=O
Appearance (Color)
White to yellow
Insoluble matter
=<0.005 % (in dilute HNO3)
Chloride (Cl)
=<0.005 %
Iron (Fe)
=<0.001 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text